accessibility menu, dialog, popup

UserName

CAS RN 1346688-54-7

IUPAC Name: 4,10-dibromo-7-octyl-3,11-dithia-7-azatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
Synonyms: 4,10-dibromo-7-octyl-3,11-dithia-7-azatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
Molecular Weight (g/mol): 449.26
Molecular Formula: C16H19Br2NS2
InChi Key: IJEKMGUFKKPWLT-UHFFFAOYSA-N
SMILES: CCCCCCCCN1C2=C(SC(Br)=C2)C2=C1C=C(Br)S2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results