accessibility menu, dialog, popup

UserName

FRb

CAS RN 13446-74-7

IUPAC Name: rubidium(1+) fluoride
Synonyms: rubidium fluoride
Molecular Weight (g/mol): 104.47
Molecular Formula: FRb
InChi Key: AHLATJUETSFVIM-UHFFFAOYSA-M
SMILES: [F-].[Rb+]
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results