accessibility menu, dialog, popup

UserName

HClH3COH2N

CAS RN 134-29-2

IUPAC Name: hydrogen 2-methoxyaniline chloride
Synonyms: hydrogen O-anisidine chloride
Molecular Weight (g/mol): 159.61
Molecular Formula: C7H10ClNO
InChi Key: XCZCWGVXRBJCCD-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=CC=CC=C1N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results