accessibility menu, dialog, popup

UserName

CAS RN 1333-39-7

OHSHOOO

CAS RN 1333-39-7

IUPAC Name: 2-hydroxybenzene-1-sulfonic acid
Synonyms: 2-hydroxybenzenesulfonic acid
Molecular Weight (g/mol): 174.17
Molecular Formula: C6H6O4S
InChi Key: IULJSGIJJZZUMF-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1S(O)(=O)=O
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)