accessibility menu, dialog, popup

UserName

CAS RN 1331742-86-9

IUPAC Name: 7-bromo-2,1,3-benzothiadiazole-4-carbonitrile
Synonyms: 7-bromo-2,1,3-benzothiadiazole-4-carbonitrile
Molecular Weight (g/mol): 240.08
Molecular Formula: C7H2BrN3S
InChi Key: ORLZWSSVSNUQAZ-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C#N)C2=NSN=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results