accessibility menu, dialog, popup

UserName

CAS RN 133005-88-6

IUPAC Name: 4-[2-(4-carboxyphenyl)ethenyl]benzoic acid
Synonyms: 4-[2-(4-carboxyphenyl)ethenyl]benzoic acid
Molecular Weight (g/mol): 268.27
Molecular Formula: C16H12O4
InChi Key: SBBQDUFLZGOASY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=CC2=CC=C(C=C2)C(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results