accessibility menu, dialog, popup

UserName

CAS RN 132765-36-7

IUPAC Name: 3-[(2-{4,5-bis[(2-cyanoethyl)sulfanyl]-2H-1,3-dithiol-2-ylidene}-5-[(2-cyanoethyl)sulfanyl]-2H-1,3-dithiol-4-yl)sulfanyl]propanenitrile
Synonyms: 3-[(2-{4,5-bis[(2-cyanoethyl)sulfanyl]-1,3-dithiol-2-ylidene}-5-[(2-cyanoethyl)sulfanyl]-1,3-dithiol-4-yl)sulfanyl]propanenitrile
Molecular Weight (g/mol): 544.83
Molecular Formula: C18H16N4S8
InChi Key: OFNXHINRTQFOKX-UHFFFAOYSA-N
SMILES: N#CCCSC1=C(SCCC#N)SC(S1)=C1SC(SCCC#N)=C(SCCC#N)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results