accessibility menu, dialog, popup

UserName

CAS RN 132071-87-5

PPClRuCl(?)

CAS RN 132071-87-5

IUPAC Name: [2'-(diphenylphosphanyl)-[1,1'-binaphthalen]-2-yl]diphenylphosphane; dichlororuthenium
Synonyms: binap; dichlororuthenium
Molecular Weight (g/mol): 794.66
Molecular Formula: C44H32Cl2P2Ru
InChi Key: YEKBVMDAGDTOQB-UHFFFAOYSA-L
SMILES: Cl[Ru]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results