accessibility menu, dialog, popup

UserName

CAS RN 132-17-2

IUPAC Name: (1R,5R)-3-(diphenylmethoxy)-8-methyl-8-azabicyclo[3.2.1]octane; methanesulfonic acid
Synonyms: benztropine; methanesulfonic acid
Molecular Weight (g/mol): 403.54
Molecular Formula: C22H29NO4S
InChi Key: CPFJLLXFNPCTDW-STYNFMPRSA-N
SMILES: CS(O)(=O)=O.CN1[C@@H]2CC[C@@H]1CC(C2)OC(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results