accessibility menu, dialog, popup

UserName

CAS RN 131894-91-2

IUPAC Name: 1-heptyl-4-iodobenzene
Synonyms: 1-heptyl-4-iodobenzene
Molecular Weight (g/mol): 302.2
Molecular Formula: C13H19I
InChi Key: NHXKOPSTJPLWPK-UHFFFAOYSA-N
SMILES: CCCCCCCC1=CC=C(I)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results