accessibility menu, dialog, popup

UserName

CAS RN 13185-00-7

IUPAC Name: 6,6'-dibromo-[1,1'-binaphthalene]-2,2'-diol
Synonyms: 6,6'-dibromo-[1,1'-binaphthalene]-2,2'-diol
Molecular Weight (g/mol): 444.12
Molecular Formula: C20H12Br2O2
InChi Key: OORIFUHRGQKYEV-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=C(Br)C=CC2=C1C1=C2C=CC(Br)=CC2=CC=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results