accessibility menu, dialog, popup

UserName

CAS RN 131818-17-2

IUPAC Name: tert-butyl N-(4-bromophenyl)carbamate
Synonyms: tert-butyl N-(4-bromophenyl)carbamate
Molecular Weight (g/mol): 272.14
Molecular Formula: C11H14BrNO2
InChi Key: VLGPDTPSKUUHKR-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results