CAS RN 131747-40-5
IUPAC Name:
5-(trifluoromethyl)pyridine-3-carboxylic acid
Synonyms:
5-(trifluoromethyl)pyridine-3-carboxylic acid
Molecular Weight (g/mol):
191.11
Molecular Formula:
C7H4F3NO2
InChi Key:
RKFDCELCLIZRRH-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(=CN=C1)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
5-(Trifluoromethyl)nicotinic acid, 97%, Thermo Scientific Chemicals
5-(Trifluoromethyl)nicotinic acid, 97%, Thermo Scientific Chemicals
CAS: 131747-40-5 Molecular Formula: C7H4F3NO2 Molecular Weight (g/mol): 191.11 MDL Number: MFCD07774128 InChI Key: RKFDCELCLIZRRH-UHFFFAOYSA-N Synonym: 5-trifluoromethyl nicotinic acid,5-trifluoromethyl pyridine-3-carboxylic acid,5-trifluoromethylinicotinic acid,5-trifluoromethyl-3-pyridinecarboxylic acid,3-pyridinecarboxylic acid, 5-trifluoromethyl,5-trifluoromethyl-nicotinic acid,3-pyridinecarboxylicacid, 5-trifluoromethyl,5-trifluoromethy pyridine-3-carboxylic acid PubChem CID: 10976305 IUPAC Name: 5-(trifluoromethyl)pyridine-3-carboxylic acid SMILES: OC(=O)C1=CC(=CN=C1)C(F)(F)F