accessibility menu, dialog, popup

UserName

H3COOOOCH3

CAS RN 13118-91-7

IUPAC Name: 1-methoxy-4-[3-(4-methoxyphenoxy)phenoxy]benzene
Synonyms: 1-methoxy-4-[3-(4-methoxyphenoxy)phenoxy]benzene
Molecular Weight (g/mol): 322.36
Molecular Formula: C20H18O4
InChi Key: MUFYVNCRQIHDCE-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC2=CC(OC3=CC=C(OC)C=C3)=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results