accessibility menu, dialog, popup

UserName

ClClClCl

CAS RN 13116-53-5

IUPAC Name: 1,2,2,3-tetrachloropropane
Synonyms: propane, 1,2,2,3-tetrachloro-
Molecular Weight (g/mol): 181.87
Molecular Formula: C3H4Cl4
InChi Key: UDPHJTAYHSSOQB-UHFFFAOYSA-N
SMILES: ClCC(Cl)(Cl)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results