accessibility menu, dialog, popup

UserName

NHNNHNH2OH2OHClHClHClHCl

CAS RN 1310544-60-5

IUPAC Name: [(pyridin-2-yl)methyl](2-{[(pyridin-2-yl)methyl]amino}ethyl)amine dihydrate tetrahydrochloride
Synonyms: (pyridin-2-ylmethyl)({2-[(pyridin-2-ylmethyl)amino]ethyl})amine dihydrate tetrahydrochloride
Molecular Weight (g/mol): 424.19
Molecular Formula: C14H26Cl4N4O2
InChi Key: OQNCOXYRPPPEIW-UHFFFAOYSA-N
SMILES: O.O.Cl.Cl.Cl.Cl.C(CNCC1=CC=CC=N1)NCC1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results