accessibility menu, dialog, popup

UserName

OHOHOHOOH

CAS RN 131-55-5

IUPAC Name: 4-(2,4-dihydroxybenzoyl)benzene-1,3-diol
Synonyms: benzophenone-2
Molecular Weight (g/mol): 246.22
Molecular Formula: C13H10O5
InChi Key: WXNRYSGJLQFHBR-UHFFFAOYSA-N
SMILES: OC1=CC=C(C(=O)C2=CC=C(O)C=C2O)C(O)=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)