CAS RN 131-54-4
IUPAC Name:
2-(2-hydroxy-4-methoxybenzoyl)-5-methoxyphenol
Synonyms:
benzophenone-6
Molecular Weight (g/mol):
274.27
Molecular Formula:
C15H14O5
InChi Key:
SODJJEXAWOSSON-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C(=O)C2=CC=C(OC)C=C2O)C(O)=C1
1
–
1
of
1
results
1
2,2'-Dihydroxy-4,4'-dimethoxybenzophenone 90.0+%, TCI America™
CAS: 131-54-4 Molecular Formula: C15H14O5 Molecular Weight (g/mol): 274.272 MDL Number: MFCD00009606 InChI Key: SODJJEXAWOSSON-UHFFFAOYSA-N Synonym: 2,2'-dihydroxy-4,4'-dimethoxybenzophenone,bis 2-hydroxy-4-methoxyphenyl methanone,benzophenone-6,cyasorb uv 12,methanone, bis 2-hydroxy-4-methoxyphenyl,uvinul d 49,caswell no. 353c,unii-7813j9cs1g,2,2-dihydroxy-4,4-dimethoxybenzophenone,4,4'-dimethoxy-2,2'-dihydroxybenzophenone PubChem CID: 8570 IUPAC Name: bis(2-hydroxy-4-methoxyphenyl)methanone SMILES: COC1=CC(=C(C=C1)C(=O)C2=C(C=C(C=C2)OC)O)O