CAS RN 13080-89-2
IUPAC Name:
4-{4-[4-(4-aminophenoxy)benzenesulfonyl]phenoxy}aniline
Synonyms:
4-{4-[4-(4-aminophenoxy)benzenesulfonyl]phenoxy}aniline
Molecular Weight (g/mol):
432.49
Molecular Formula:
C24H20N2O4S
InChi Key:
UTDAGHZGKXPRQI-UHFFFAOYSA-N
SMILES:
NC1=CC=C(OC2=CC=C(C=C2)S(=O)(=O)C2=CC=C(OC3=CC=C(N)C=C3)C=C2)C=C1
1
–
1
of
1
results
1
Bis[4-(4-aminophenoxy)phenyl] Sulfone 98.0+%, TCI America™
CAS: 13080-89-2 Molecular Formula: C24H20N2O4S Molecular Weight (g/mol): 432.49 MDL Number: MFCD00039155 InChI Key: UTDAGHZGKXPRQI-UHFFFAOYSA-N PubChem CID: 25689 IUPAC Name: 4-{4-[4-(4-aminophenoxy)benzenesulfonyl]phenoxy}aniline SMILES: NC1=CC=C(OC2=CC=C(C=C2)S(=O)(=O)C2=CC=C(OC3=CC=C(N)C=C3)C=C2)C=C1