accessibility menu, dialog, popup

UserName

CAS RN 13050-47-0

IUPAC Name: 3-methylbenzohydrazide
Synonyms: benzoylhydrazine, M-methyl
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: XFNNAMBYJSQXKF-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C(=O)NN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results