accessibility menu, dialog, popup

UserName

H3CCH2

CAS RN 129738-34-7

IUPAC Name: 4-ethenyl-4'-pentyl-1,1'-bi(cyclohexane)
Synonyms: 4-ethenyl-4'-pentyl-1,1'-bi(cyclohexane)
Molecular Weight (g/mol): 262.48
Molecular Formula: C19H34
InChi Key: ALEUVXQGVOQDBD-UHFFFAOYSA-N
SMILES: CCCCCC1CCC(CC1)C1CCC(CC1)C=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results