accessibility menu, dialog, popup

UserName

CAS RN 129575-89-9

IUPAC Name: 2-[4-(benzyloxy)-6-[(benzyloxy)methyl]-5-hydroxy-2-(4-methoxyphenoxy)oxan-3-yl]-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-[4-(benzyloxy)-6-[(benzyloxy)methyl]-5-hydroxy-2-(4-methoxyphenoxy)oxan-3-yl]isoindole-1,3-dione
Molecular Weight (g/mol): 595.65
Molecular Formula: C35H33NO8
InChi Key: QKFDXGIKVGVBGT-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(COCC3=CC=CC=C3)C(O)C(OCC3=CC=CC=C3)C2N2C(=O)C3=CC=CC=C3C2=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results