accessibility menu, dialog, popup

UserName

H3COSSBrOCH3Br

CAS RN 1294515-75-5

IUPAC Name: 5,11-dibromo-2,8-bis(octyloxy)-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene
Synonyms: 5,11-dibromo-2,8-bis(octyloxy)-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene
Molecular Weight (g/mol): 604.5
Molecular Formula: C26H36Br2O2S2
InChi Key: GXGDTYKIKNMZTM-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=C2C=C(Br)SC2=C(OCCCCCCCC)C2=C1SC(Br)=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results