accessibility menu, dialog, popup

UserName

OOSCl

CAS RN 129-94-2

IUPAC Name: 2-[(8-chloronaphthalen-1-yl)sulfanyl]acetate
Synonyms: 2-[(8-chloronaphthalen-1-yl)sulfanyl]acetate
Molecular Weight (g/mol): 251.7
Molecular Formula: C12H8ClO2S
InChi Key: WPNAGQJVWAFQJV-UHFFFAOYSA-M
SMILES: [O-]C(=O)CSC1=C2C(Cl)=CC=CC2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results