accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHOOCH3CH3H3C

CAS RN 128988-04-5

IUPAC Name: tert-butyl 3-{[3-(tert-butoxy)-3-oxopropyl]amino}propanoate
Synonyms: tert-butyl 3-{[3-(tert-butoxy)-3-oxopropyl]amino}propanoate
Molecular Weight (g/mol): 273.37
Molecular Formula: C14H27NO4
InChi Key: KCTOWZYKZDFZMQ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CCNCCC(=O)OC(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)