accessibility menu, dialog, popup

UserName

OHOHOOHOOOHO

CAS RN 128-97-2

IUPAC Name: naphthalene-1,4,5,8-tetracarboxylic acid
Synonyms: naphthalene-1,4,5,8-tetracarboxylic acid
Molecular Weight (g/mol): 304.21
Molecular Formula: C14H8O8
InChi Key: OLAPPGSPBNVTRF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C(O)=O)C2=C(C=CC(C(O)=O)=C12)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results