accessibility menu, dialog, popup

UserName

CAS RN 127337-60-4

IUPAC Name: 2-(chloromethyl)-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine hydrochloride
Synonyms: 2-(chloromethyl)-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine hydrochloride
Molecular Weight (g/mol): 276.08
Molecular Formula: C9H10Cl2F3NO
InChi Key: CMZBQUWICURDCD-UHFFFAOYSA-N
SMILES: Cl.CC1=C(OCC(F)(F)F)C=CN=C1CCl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results