accessibility menu, dialog, popup

UserName

NaNH2AsHOOO

CAS RN 127-85-5

IUPAC Name: sodium hydrogen (4-aminophenyl)arsonate
Synonyms: atoxyl sodium anilarsonate arsanilic acid sodium salt sodium aminarsonate sodium p-aminophenylarsonate sodium arsanilate arsamin
Molecular Weight (g/mol): 239.04
Molecular Formula: C6H7AsNNaO3
InChi Key: OUFRIWNNMFWZTM-UHFFFAOYSA-M
SMILES: [Na+].NC1=CC=C(C=C1)[As](O)([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results