accessibility menu, dialog, popup

UserName

OOOONi

CAS RN 126956-48-7

IUPAC Name: nickel(2+) oxalate
Synonyms: nickel oxalate
Molecular Weight (g/mol): 146.71
Molecular Formula: C2NiO4
InChi Key: DOLZKNFSRCEOFV-UHFFFAOYSA-L
SMILES: [Ni++].[O-]C(=O)C([O-])=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)