accessibility menu, dialog, popup

UserName

CAS RN 126716-90-3

OOHOHO

CAS RN 126716-90-3

IUPAC Name: 4-[3-(4-hydroxyphenoxy)phenoxy]phenol
Synonyms: 4-[3-(4-hydroxyphenoxy)phenoxy]phenol
Molecular Weight (g/mol): 294.31
Molecular Formula: C18H14O4
InChi Key: CJLPIPXJJJUBIV-UHFFFAOYSA-N
SMILES: OC1=CC=C(OC2=CC(OC3=CC=C(O)C=C3)=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results