accessibility menu, dialog, popup

UserName

CAS RN 126-96-5

NaCH3OHOCH3OO

CAS RN 126-96-5

IUPAC Name: sodium acetic acid acetate
Synonyms: sodium diacetate
Molecular Weight (g/mol): 142.09
Molecular Formula: C4H7NaO4
InChi Key: BHZOKUMUHVTPBX-UHFFFAOYSA-M
SMILES: [Na+].CC(O)=O.CC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results