accessibility menu, dialog, popup

UserName

CAS RN 1256355-57-3

IUPAC Name: (5-cyano-2-hydroxyphenyl)boronic acid
Synonyms: 5-cyano-2-hydroxyphenylboronic acid
Molecular Weight (g/mol): 162.94
Molecular Formula: C7H6BNO3
InChi Key: QXWBHAJKEPWEBD-UHFFFAOYSA-N
SMILES: OB(O)C1=C(O)C=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results