CAS RN 1256259-14-9
CAS RN 1256259-14-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(Bromomethyl)-7-(diethylamino)coumarin 98.0+%, TCI America™
CAS: 1256259-14-9 Molecular Formula: C14H16BrNO2