CAS RN 125455-64-3
IUPAC Name:
N-(2-{[3-acetamido-5-hydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)oxan-4-yl]oxy}-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl)acetamide
Synonyms:
N-(2-{[3-acetamido-5-hydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)oxan-4-yl]oxy}-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl)acetamide
Molecular Weight (g/mol):
545.5
Molecular Formula:
C22H31N3O13
InChi Key:
HXQAUFSCNOLKJP-UHFFFAOYNA-N
SMILES:
CC(=O)NC1C(O)C(O)C(CO)OC1OC1C(O)C(CO)OC(OC2=CC=C(C=C2)[N+]([O-])=O)C1NC(C)=O
1
–
1
of
1
results
1
GlcNAc beta(1-3)GalNAc-alpha-pNP 98.0+%, TCI America™
CAS: 125455-64-3 Molecular Formula: C22H31N3O13 Molecular Weight (g/mol): 545.498 InChI Key: HXQAUFSCNOLKJP-GVDFRNMQSA-N PubChem CID: 72943121 IUPAC Name: N-[(3R,4R,5S,6R)-2-[(2R,3R,4R,5R,6R)-5-acetamido-3-hydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-4-yl]oxy-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide SMILES: CC(=O)NC1C(C(C(OC1OC2C(C(OC(C2O)CO)OC3=CC=C(C=C3)[N+](=O)[O-])NC(=O)C)CO)O)O