accessibility menu, dialog, popup

UserName

CAS RN 124-47-0

IUPAC Name: nitric acid; urea
Synonyms: urea nitrate
Molecular Weight (g/mol): 123.07
Molecular Formula: CH5N3O4
InChi Key: AYTGUZPQPXGYFS-UHFFFAOYSA-N
SMILES: NC(N)=O.O[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results