accessibility menu, dialog, popup

UserName

BrI

CAS RN 123348-27-6

IUPAC Name: 2-bromo-7-iodo-9H-fluorene
Synonyms: 2-bromo-7-iodo-9H-fluorene
Molecular Weight (g/mol): 371.02
Molecular Formula: C13H8BrI
InChi Key: GWUPMAMGLLLLHO-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)C1=C(C2)C=C(I)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results