accessibility menu, dialog, popup

UserName

CAS RN 1229444-44-3

H3CNPBr

CAS RN 1229444-44-3

IUPAC Name: 1-{[4-(diphenylphosphanyl)phenyl]methyl}-1-methylpyrrolidin-1-ium bromide
Synonyms: 1-{[4-(diphenylphosphanyl)phenyl]methyl}-1-methylpyrrolidin-1-ium bromide
Molecular Weight (g/mol): 440.37
Molecular Formula: C24H27BrNP
InChi Key: JKJSVXHLMBCNPA-UHFFFAOYSA-M
SMILES: [Br-].C[N+]1(CC2=CC=C(C=C2)P(C2=CC=CC=C2)C2=CC=CC=C2)CCCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results