CAS RN 1228185-09-8
IUPAC Name:
1,3-bis[2,6-bis(propan-2-yl)phenyl]-2-chloro-3H-1λ⁵-imidazol-1-ylium chloride
Synonyms:
2-chloro-1,3-bis(2,6-diisopropylphenyl)-1λ⁵-imidazol-1-ylium chloride
Molecular Weight (g/mol):
459.5
Molecular Formula:
C27H36Cl2N2
InChi Key:
JDMACANGISWEGX-UHFFFAOYSA-M
SMILES:
[Cl-].CC(C)C1=CC=CC(C(C)C)=C1N1C=C[N+](=C1Cl)C1=C(C=CC=C1C(C)C)C(C)C
1
–
1
of
1
results
1
2-Chloro-1,3-bis(2,6-diisopropylphenyl)-1H-imidazolium Chloride 98.0+%, TCI America™
CAS: 1228185-09-8 Molecular Formula: C27H36Cl2N2 Molecular Weight (g/mol): 459.50 MDL Number: MFCD23703069 InChI Key: JDMACANGISWEGX-UHFFFAOYSA-M PubChem CID: 71463958 IUPAC Name: 1,3-bis[2,6-bis(propan-2-yl)phenyl]-2-chloro-3H-1λ⁵-imidazol-1-ylium chloride SMILES: [Cl-].CC(C)C1=CC=CC(C(C)C)=C1N1C=C[N+](=C1Cl)C1=C(C=CC=C1C(C)C)C(C)C