accessibility menu, dialog, popup

UserName

HOONN

CAS RN 122773-97-1

IUPAC Name: 2-phenylpyrimidine-5-carboxylic acid
Synonyms: 2-phenylpyrimidine-5-carboxylic acid
Molecular Weight (g/mol): 200.2
Molecular Formula: C11H8N2O2
InChi Key: BOAIYSRFGWBZCF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(N=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)