accessibility menu, dialog, popup

UserName

ClClIrIr(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 12246-51-4

IUPAC Name: tetrakis(cyclooctene) diiridium dichloride
Synonyms: chlorobis(cyclooctene)iridium dimer
Molecular Weight (g/mol): 896.14
Molecular Formula: C32H56Cl2Ir2
InChi Key: CJJIQMGSHWWMCK-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Ir].[Ir].C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results