accessibility menu, dialog, popup

UserName

CAS RN 12145-47-0

IUPAC Name: (1Z,5Z)-cycloocta-1,5-diene; dibromopalladiumylium
Synonyms: 1,5-cyclooctadiene, (Z,Z)-; dibromopalladiumylium
Molecular Weight (g/mol): 374.41
Molecular Formula: C8H12Br2Pd
InChi Key: MTEMVOIUTLIPJT-PHFPKPIQSA-L
SMILES: Br[Pd+]Br.C1C\C=C/CC\C=C/1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results