accessibility menu, dialog, popup

UserName

CAS RN 1214386-29-4

IUPAC Name: 4-bromo-2-chloro-5-fluorobenzaldehyde
Synonyms: 4-bromo-2-chloro-5-fluorobenzaldehyde
Molecular Weight (g/mol): 237.45
Molecular Formula: C7H3BrClFO
InChi Key: XPEGZWLIWARWAA-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=C(Cl)C(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results