accessibility menu, dialog, popup

UserName

CAS RN 121219-07-6

CH3OFF

CAS RN 121219-07-6

IUPAC Name: 1-ethoxy-2,3-difluorobenzene
Synonyms: 1-ethoxy-2,3-difluorobenzene
Molecular Weight (g/mol): 158.15
Molecular Formula: C8H8F2O
InChi Key: AVOGLGBKOFOSBN-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results