accessibility menu, dialog, popup

UserName

CH3ONHSNH2OO

CAS RN 121-61-9

IUPAC Name: N-(4-sulfamoylphenyl)acetamide
Synonyms: acetylsulfanilamide
Molecular Weight (g/mol): 214.24
Molecular Formula: C8H10N2O3S
InChi Key: PKOFBDHYTMYVGJ-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)S(N)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results