accessibility menu, dialog, popup

UserName

CH3OHOFFF

CAS RN 120985-64-0

IUPAC Name: 4-methyl-2-(trifluoromethyl)benzoic acid
Synonyms: 4-methyl-2-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: PHJOGJTZXVBKJO-UHFFFAOYSA-N
SMILES: CC1=CC(=C(C=C1)C(O)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results