CAS RN 12059-14-2
IUPAC Name:
dinickeliosilane
Synonyms:
dinickeliosilane
Molecular Weight (g/mol):
145.47
Molecular Formula:
Ni2Si
InChi Key:
RUFLMLWJRZAWLJ-UHFFFAOYSA-N
SMILES:
[Ni]=[Si]=[Ni]
1
–
1
of
1
results
1
Nickel silicide, 99% (metals basis excluding Co), Co 0.1-1%, Thermo Scientific Chemicals
CAS: 12059-14-2 Molecular Formula: Ni2Si Molecular Weight (g/mol): 145.472 MDL Number: MFCD00151367 InChI Key: RUFLMLWJRZAWLJ-UHFFFAOYSA-N Synonym: nickel silicide,dinickelylidenesilane PubChem CID: 14767304 SMILES: [Si](=[Ni])=[Ni]