accessibility menu, dialog, popup

UserName

CAS RN 12057-71-5

NNMgMgMg

CAS RN 12057-71-5

IUPAC Name: diazenediide; trimagnesium-1,3-bis(ylium)
Synonyms: magnesium nitride
Molecular Weight (g/mol): 100.93
Molecular Formula: Mg3N2
InChi Key: VZPQZBKNAUHPOS-UHFFFAOYSA-N
SMILES: [N-]=[N-].[Mg+][Mg][Mg+]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results