accessibility menu, dialog, popup

UserName

OSOOOPtNNOOOOH3OH3O

CAS RN 12033-81-7

IUPAC Name: dihydrogen dinitroplatinumbis(ylium) sulfate
Synonyms: dihydrogen dinitroplatinumbis(ylium) sulfate
Molecular Weight (g/mol): 385.16
Molecular Formula: H2N2O8PtS
InChi Key: SPPCQWREVYSZTJ-UHFFFAOYSA-N
SMILES: [H+].[H+].[O-]S([O-])(=O)=O.[O-][N+](=O)[Pt++][N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results