accessibility menu, dialog, popup

UserName

OHOOOOOOHMg

CAS RN 119851-23-9

IUPAC Name: magnesium(2+) 2-(carboxymethyl)-2-hydroxybutanedioate
Synonyms: magnesium citrate
Molecular Weight (g/mol): 214.41
Molecular Formula: C6H6MgO7
InChi Key: DIXGJWCZQHXZNR-UHFFFAOYNA-L
SMILES: [Mg++].OC(=O)CC(O)(CC([O-])=O)C([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results