accessibility menu, dialog, popup

UserName

H3CHNCH3

CAS RN 1198395-24-2

IUPAC Name: N-{[1,1'-biphenyl]-2-yl}-9,9-dimethyl-9H-fluoren-2-amine
Synonyms: N-{[1,1'-biphenyl]-2-yl}-9,9-dimethylfluoren-2-amine
Molecular Weight (g/mol): 361.49
Molecular Formula: C27H23N
InChi Key: OBARUOOPPWHZRQ-UHFFFAOYSA-N
SMILES: CC1(C)C2=CC=CC=C2C2=C1C=C(NC1=CC=CC=C1C1=CC=CC=C1)C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results